a-Tosyl(2-bromobenzyl)isocyanide ≥95%

Supplier: Matrix Scientific

Synonyms: a-Tosyl-(2-bromobenzyl) isocyanide

071798-1G 071798-5G 071798-10G
102521-394EA 533.85 USD
102521-394 102521-396 102521-398
a-Tosyl(2-bromobenzyl)isocyanide ≥95%
a-Tosyl(2-bromobenzyl)isocyanide

Hazard: Irritant

Formula: C₁₅H₁₂BrNO₂S
MW: 350.2303 g/mol
MDL Number: MFCD04114784
CAS Number: 936548-16-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR