5-(p-Tolyl)pyrimidin-2-amine ≥95%

Supplier: Matrix Scientific

071795-1G 071795-5G
102521-380EA 355.38 USD
102521-380 102521-382
5-(p-Tolyl)pyrimidin-2-amine ≥95%
5-(p-Tolyl)pyrimidin-2-amine

Hazard: Irritant

Formula: C₁₁H₁₁N₃
CAS Number: 31408-17-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR