2-Aminopyrimidine-5-carbonitrile ≥95%

Supplier: Matrix Scientific

071792-1G 071792-5G
102521-362EA 84.62 USD
102521-362 102521-364
2-Aminopyrimidine-5-carbonitrile ≥95%
2-Aminopyrimidine-5-carbonitrile

Hazard: Irritant

Formula: C₅H₄N₄
MW: 120.1121 g/mol
MDL Number: MFCD00232791
CAS Number: 1753-48-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR