1-Boc-3-bromo-7-nitroindole ≥95%

Supplier: Matrix Scientific

Synonyms: 3-Bromo-7-nitroindole, N-BOC protected, 1-Boc-3-Bromo-7-nitroindole

071791-1G 071791-5G 071791-10G
102521-356EA 355.38 USD
102521-356 102521-358 102521-360
1-Boc-3-bromo-7-nitroindole ≥95%
1-Boc-3-bromo-7-nitroindole

Hazard: Irritant

Formula: C₁₃H₁₃BrN₂O₄
MW: 341.1600 g/mol
MDL Number: MFCD05864785
CAS Number: 914349-37-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR