1-Boc-3-bromoindole ≥95%

Supplier: Matrix Scientific
Warning

Synonyms: tert-Butyl 3-bromo-1H-indole-1-carboxylate, tert-Butyl-3-bromo-1H-indole-1-carboxylate, 3-Bromoindole-1-carboxylic acid tert-butyl ester

071790-1G 071790-10G
102521-350EA 38.46 USD
102521-350 102521-354
1-Boc-3-bromoindole ≥95%
1-Boc-3-bromoindole

Purity: 95+% CAS: 143259-56-7 MDL Number: MFCD05864772 Molecular Formula: C13H14BrNO2 Molecular Weight: 296,17 Hazard: Irritant

Formula: C₁₃H₁₄BrNO₂
MW: 296.16 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD05864772
CAS Number: 143259-56-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR