1-Boc-3-bromo-5-methoxyindole ≥95%

Supplier: Matrix Scientific

071789-1G 071789-5G 071789-10G
102521-344EA 249.23 USD
102521-344 102521-346 102521-348
1-Boc-3-bromo-5-methoxyindole ≥95%
1-Boc-3-bromo-5-methoxyindole

Hazard: Irritant

Formula: C₁₄H₁₆BrNO₃
CAS Number: 348640-11-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR