(1-Pyridin-2-ylmethylpiperidin-4-yl)methanol ≥95%

Supplier: Matrix Scientific

071788-1G 071788-5G 071788-10G
102521-338EA 355.38 USD
102521-338 102521-340 102521-342
(1-Pyridin-2-ylmethylpiperidin-4-yl)methanol ≥95%
(1-Pyridin-2-ylmethylpiperidin-4-yl)methanol

Hazard: Irritant

Formula: C₁₂H₁₈N₂O
CAS Number: 914349-21-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR