1-(Pyridin-2-ylmethyl)piperidine-4-carboxylic acid ≥95%

Supplier: Matrix Scientific

071786-1G 071786-5G 071786-10G
102521-326EA 355.38 USD
102521-326 102521-328 102521-330
1-(Pyridin-2-ylmethyl)piperidine-4-carboxylic acid ≥95%
1-(Pyridin-2-ylmethyl)piperidine-4-carboxylic acid

Hazard: Irritant

Formula: C₁₂H₁₆N₂O₂
MW: 220.2700 g/mol
MDL Number: MFCD03371469
CAS Number: 193538-28-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR