(4-Phenylpiperazin-1-yl)(piperidin-4-yl)methanone dihydrochloride ≥95%

Supplier: Matrix Scientific

071781-1G 071781-5G 071781-10G
102521-296EA 284.62 USD
102521-296 102521-298 102521-300
(4-Phenylpiperazin-1-yl)(piperidin-4-yl)methanone dihydrochloride ≥95%
(4-Phenylpiperazin-1-yl)(piperidin-4-yl)methanone dihydrochloride

Hazard: Irritant

Formula: C₁₆H₂₅Cl₂N₃O
CAS Number: 1184996-07-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR