(4-Methylpiperazin-1-yl)piperidin-4-ylmethanone dihydrochloride ≥95%

Supplier: Matrix Scientific

071778-1G 071778-5G 071778-10G
102521-278EA 213.85 USD
102521-278 102521-280 102521-282
(4-Methylpiperazin-1-yl)piperidin-4-ylmethanone dihydrochloride ≥95%
(4-Methylpiperazin-1-yl)piperidin-4-ylmethanone dihydrochloride

Hazard: Irritant

Formula: C₁₁H₂₃Cl₂N₃O
MDL Number: MFCD05864745
CAS Number: 1019852-03-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR