2-(Piperidin-4-yloxy)pyrimidine ≥95%

Supplier: Matrix Scientific

071777-1G 071777-5G 071777-10G
102521-272EA 167.69 USD
102521-272 102521-274 102521-276
2-(Piperidin-4-yloxy)pyrimidine ≥95%
2-(Piperidin-4-yloxy)pyrimidine

Hazard: Irritant

Formula: C₉H₁₃N₃O
CAS Number: 499240-48-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR