1-Pyridin-2-ylmethylpiperidin-4-one ≥95%

Supplier: Matrix Scientific

071776-1G 071776-5G 071776-10G
102521-266EA 284.62 USD
102521-266 102521-268 102521-270
1-Pyridin-2-ylmethylpiperidin-4-one ≥95%
1-Pyridin-2-ylmethylpiperidin-4-one

Hazard: Irritant

Formula: C₁₁H₁₄N₂O
MW: 190.2417 g/mol
MDL Number: MFCD05864492
CAS Number: 41661-56-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR