1-(Pyrimidin-2-yl)piperidin-4-ol ≥95%

Supplier: Matrix Scientific

071775-1G 071775-5G 071775-10G
102521-260EA 284.62 USD
102521-260 102521-262 102521-264
1-(Pyrimidin-2-yl)piperidin-4-ol ≥95%
1-(Pyrimidin-2-yl)piperidin-4-ol

Hazard: Irritant

Formula: C₉H₁₃N₃O
MW: 179.2200 g/mol
MDL Number: MFCD05864742
CAS Number: 893755-98-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR