N-Boc-4-[2-(4-Toluenesulfonyloxy)ethyl]piperidine ≥95%

Supplier: Matrix Scientific

Synonyms: tert-Butyl 4-(2-(tosyloxy)ethyl)piperidine-1-carboxylate

071774-1G 071774-5G 071774-10G
102521-254EA 355.38 USD
102521-254 102521-256 102521-258
N-Boc-4-[2-(4-Toluenesulfonyloxy)ethyl]piperidine ≥95%
N-Boc-4-[2-(4-Toluenesulfonyloxy)ethyl]piperidine

Hazard: Irritant

Formula: C₁₉H₂₉NO₅S
MW: 383.502 g/mol
MDL Number: MFCD05864741
CAS Number: 89151-45-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR