N-Cbz-2-piperidinecarboxylic acid ≥95%

Supplier: Matrix Scientific

071773-5G 071773-100G 071773-25G
102521-248EA 15.38 USD
102521-248 102521-252 102833-508
N-Cbz-2-piperidinecarboxylic acid ≥95%
N-Cbz-2-piperidinecarboxylic acid

Hazard: Irritant

Formula: C₁₄H₁₇NO₄
MDL Number: MFCD01863568
CAS Number: 28697-07-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR