1-Boc-4-(6-chloropyridazin-3-yl)piperazine ≥95%
Supplier: Matrix Scientific
Synonyms:
1-Boc-4-(6-Chloropyridazin-3-yl)piperazine
071771-1G
071771-5G
071771-10G
102521-236EA
100
USD
102521-236
102521-238
102521-240
1-Boc-4-(6-chloropyridazin-3-yl)piperazine ≥95%
1-Boc-4-(6-chloropyridazin-3-yl)piperazine
Hazard: Irritant
Formula:
C₁₃H₁₉ClN₄O₂ MW: 298.7700 g/mol |
MDL Number:
MFCD05864738 CAS Number: 492431-11-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...