3-Chloro-6-(pyrrol-1-yl)pyridazine ≥95%

Supplier: Matrix Scientific

071769-1G 071769-5G 071769-10G
102521-224EA 284.62 USD
102521-224 102521-226 102521-228
3-Chloro-6-(pyrrol-1-yl)pyridazine ≥95%
3-Chloro-6-(pyrrol-1-yl)pyridazine

Hazard: Irritant

Formula: C₈H₆ClN₃
MDL Number: MFCD00828665
CAS Number: 5096-76-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR