3-Bromobenzamidine hydrochloride ≥95%

Supplier: Matrix Scientific

071767-1G 071767-5G
102521-216EA 52.31 USD
102521-216 102521-218
3-Bromobenzamidine hydrochloride ≥95%
3-Bromobenzamidine hydrochloride

Hazard: Irritant

Formula: C₇H₈BrClN₂
MDL Number: MFCD04114439
CAS Number: 16796-52-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR