3-Fluoro-N-methylbenzylamine ≥95%

Supplier: Matrix Scientific

Synonyms: (3-Fluorobenzyl)methylamine, 1-(3-Fluorophenyl)-N-methylmethanamine

071765-1G 071765-5G 071765-10G
102521-204EA 32.31 USD
102521-204 102521-206 102521-208
3-Fluoro-N-methylbenzylamine ≥95%
3-Fluoro-N-methylbenzylamine

Hazard: Irritant

Formula: C₈H₁₀FN
MW: 139.17 g/mol
MDL Number: MFCD04623547
CAS Number: 90389-84-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR