2-Fluoro-N-methylbenzylamine ≥95%

Supplier: Matrix Scientific

Synonyms: 1-(2-Fluorophenyl)-N-methylmethanamine, (2-Fluorobenzyl)methylamine

071764-5G 071764-10G
102521-200EA 613.85 USD
102521-200 102521-202
2-Fluoro-N-methylbenzylamine ≥95%
2-Fluoro-N-methylbenzylamine

Hazard: Irritant

Formula: C₈H₁₀FN
MW: 139.17 g/mol
MDL Number: MFCD04629633
CAS Number: 399-30-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR