1-(4-Bromophenyl)-N-methylmethanamine ≥95%

Supplier: Matrix Scientific

071762-1G 071762-5G 071762-10G
102521-186EA 107.69 USD
102521-186 102521-188 102521-190
1-(4-Bromophenyl)-N-methylmethanamine ≥95%
1-(4-Bromophenyl)-N-methylmethanamine

Hazard: Irritant

Formula: C₈H₁₀BrN
MDL Number: MFCD04632143
CAS Number: 699-03-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR