(2,6-Dichlorobenzyl)methylamine hydrochloride ≥95%

Supplier: Matrix Scientific

Synonyms: 1-(2,6-Dichlorophenyl)-N-methylmethanamine hydrochloride

071761-1G 071761-5G 071761-10G
102521-180EA 249.23 USD
102521-180 102521-182 102521-184
(2,6-Dichlorobenzyl)methylamine hydrochloride ≥95%
(2,6-Dichlorobenzyl)methylamine hydrochloride

Hazard: Irritant

Formula: C₈H₁₀Cl₃N
MW: 226.5307 g/mol
MDL Number: MFCD05864732
CAS Number: 90389-15-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR