(2,4-Dichlorobenzyl)methylamine ≥95%

Supplier: Matrix Scientific

Synonyms: 2,4-Dichloro-N-methylbenzylamine, N-Methyl-2,4-dichlorobenzylamine

071760-1G 071760-5G 071760-10G
102521-174EA 249.23 USD
102521-174 102521-176 102521-178
(2,4-Dichlorobenzyl)methylamine ≥95%
(2,4-Dichlorobenzyl)methylamine

Hazard: Irritant

Formula: C₈H₉Cl₂N
MW: 190.07 g/mol
MDL Number: MFCD00045185
CAS Number: 5013-77-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR