(2,3-Dichlorobenzyl)methylamine ≥95%

Supplier: Matrix Scientific

Synonyms: 1-(2,3-Dichlorophenyl)-N-methylmethanamine

071759-1G 071759-5G 071759-10G
102521-168EA 249.23 USD
102521-168 102521-170 102521-172
(2,3-Dichlorobenzyl)methylamine ≥95%
(2,3-Dichlorobenzyl)methylamine

Hazard: Irritant

Formula: C₈H₉Cl₂N
MW: 190.698 g/mol
MDL Number: MFCD04632069
CAS Number: 731827-07-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR