1-(3,4-Dichlorophenyl)-N-methylmethanamine ≥95%

Supplier: Matrix Scientific

071758-1G 071758-5G 071758-10G
102521-162EA 46.15 USD
102521-162 102521-164 102521-166
1-(3,4-Dichlorophenyl)-N-methylmethanamine ≥95%
1-(3,4-Dichlorophenyl)-N-methylmethanamine

Hazard: Irritant

Formula: C₈H₉Cl₂N
CAS Number: 5635-67-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR