4-Methoxy-N-methylbenzylamine ≥95%

Supplier: Matrix Scientific

071757-5G 071757-25G
102521-158EA 104.62 USD
102521-158 102551-784
4-Methoxy-N-methylbenzylamine ≥95%
4-Methoxy-N-methylbenzylamine

Hazard: Corrosive/Irritant

Formula: C₉H₁₃NO
MW: 151.21 g/mol
Boiling Pt: 88…90 °C (1 mmHg)
MDL Number: MFCD04115407
CAS Number: 702-24-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR