4-Methoxy-N-methylbenzylamine hydrochloride ≥95%

Supplier: Matrix Scientific

071756-1G 071756-5G 071756-10G
102521-150EA 107.69 USD
102521-150 102521-152 102521-154
4-Methoxy-N-methylbenzylamine hydrochloride ≥95%
4-Methoxy-N-methylbenzylamine hydrochloride

Hazard: Irritant

Formula: C₉H₁₄ClNO
MW: 187.6700 g/mol
MDL Number: MFCD04117881
CAS Number: 876-32-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR