tert-Butyl-4-hydroxyphenethylcarbamate ≥95%

Supplier: Matrix Scientific

Synonyms: N-Boc-4-hydroxyphenethylamine, tert-Butyl 4-hydroxyphenethylcarbamate, N-(tert-Butoxycarbonyl)-4-hydroxyphenethylamine, N-Boc-tyramine, N-(tert-Butoxycarbonyl)tyramine

071754-1G 071754-5G 071754-10G
102521-138EA 46.15 USD
102521-138 102521-140 102521-142
tert-Butyl-4-hydroxyphenethylcarbamate ≥95%
tert-Butyl-4-hydroxyphenethylcarbamate

Hazard: Irritant

Formula: C₁₃H₁₉NO₃
MDL Number: MFCD05864730
CAS Number: 64318-28-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR