Di-tert-butyl iminodicarboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Di-(tert-butyl) imidodicarbonate

071753-100G 071753-25G
102521-136EA 449.23 USD
102521-136 102663-866
Di-tert-butyl iminodicarboxylate ≥95%
Di-tert-butyl iminodicarboxylate

Hazard: Irritant

Formula: C₁₀H₁₉NO₄
MW: 217.27 g/mol
Melting Pt: 114…119 °C
MDL Number: MFCD00043309
CAS Number: 51779-32-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR