2-Amino-4-nitrobenzenemethanol ≥95%

Supplier: Matrix Scientific

071749-1G 071749-5G 071749-10G
102521-110EA 75.38 USD
102521-110 102521-112 102521-114
2-Amino-4-nitrobenzenemethanol ≥95%
2-Amino-4-nitrobenzenemethanol

Hazard: Irritant

Formula: C₇H₈N₂O₃
MDL Number: MFCD01664410
CAS Number: 78468-34-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR