(R)-1-(Pyrrolidin-2-ylmethyl)pyrrolidine ≥95%

Supplier: Matrix Scientific

Synonyms: (R)-(-)-1-(2-Pyrrolidinylmethyl)pyrrolidine

071748-1G 071748-5G 071748-10G
102521-104EA 533.85 USD
102521-104 102521-106 102521-108
(R)-1-(Pyrrolidin-2-ylmethyl)pyrrolidine ≥95%
(R)-1-(Pyrrolidin-2-ylmethyl)pyrrolidine

Hazard: Irritant

Formula: C₉H₁₈N₂
MW: 154.2500 g/mol
MDL Number: MFCD04117879
CAS Number: 60419-23-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR