Sirtinol ≥95%

Supplier: Matrix Scientific

071747-250MG 071747-1G
102521-100EA 461.54 USD
102521-100 102521-102
Sirtinol ≥95%
Sirtinol

Hazard: Irritant

Formula: C₂₆H₂₂N₂O₂
MDL Number: MFCD00810186
CAS Number: 410536-97-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR