2,6-Dichloro-3-nitrobenzaldehyde ≥95%

Supplier: Matrix Scientific

071745-1G 071745-5G 071745-10G
102521-090EA 161.54 USD
102521-090 102521-092 102521-094
2,6-Dichloro-3-nitrobenzaldehyde ≥95%
2,6-Dichloro-3-nitrobenzaldehyde

Hazard: Irritant

Formula: C₇H₃Cl₂NO₃
MDL Number: MFCD00090332
CAS Number: 5866-97-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR