1-Boc-7-benzyloxy-3-hydroxymethylindole ≥95%

Supplier: Matrix Scientific

Synonyms: 7-(Benzyloxy)-3-(hydroxymethyl)-1H-indole, N-BOC protected

071743-1G 071743-5G 071743-10G
102521-078EA 355.38 USD
102521-078 102521-080 102521-082
1-Boc-7-benzyloxy-3-hydroxymethylindole ≥95%
1-Boc-7-benzyloxy-3-hydroxymethylindole

Hazard: Irritant

Formula: C₂₁H₂₃NO₄
MW: 353.42 g/mol
MDL Number: MFCD05864726
CAS Number: 914349-16-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR