tert-Butyl-3-(hydroxymethyl)-7-nitro-1H-indole-1-carboxylate ≥95%

Supplier: Matrix Scientific

071742-1G 071742-5G 071742-10G
102521-072EA 355.38 USD
102521-072 102521-074 102521-076
tert-Butyl-3-(hydroxymethyl)-7-nitro-1H-indole-1-carboxylate ≥95%
tert-Butyl-3-(hydroxymethyl)-7-nitro-1H-indole-1-carboxylate

Hazard: Irritant

Formula: C₁₄H₁₆N₂O₅
MDL Number: MFCD05864725
CAS Number: 914349-15-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR