tert-Butyl-5-(benzyloxy)-3-(hydroxymethyl)-1H-indole-1-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: 5-(Benzyloxy)-3-(hydroxymethyl)-1H-indole, N-BOC protected

071741-1G 071741-5G 071741-10G
102521-066EA 178.46 USD
102521-066 102521-068 102521-070
tert-Butyl-5-(benzyloxy)-3-(hydroxymethyl)-1H-indole-1-carboxylate ≥95%
tert-Butyl-5-(benzyloxy)-3-(hydroxymethyl)-1H-indole-1-carboxylate

Hazard: Irritant

Formula: C₂₁H₂₃NO₄
CAS Number: 914349-14-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR