tert-Butyl-3-(hydroxymethyl)-2-methyl-1H-indole-1-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: tert-Butyl 3-(hydroxymethyl)-2-methyl-1H-indole-1-carboxylate, 3-(Hydroxymethyl)-2-methyl-1H-indole, N-BOC protected

071740-1G 071740-5G 071740-10G
102521-060EA 178.46 USD
102521-060 102521-062 102521-064
tert-Butyl-3-(hydroxymethyl)-2-methyl-1H-indole-1-carboxylate ≥95%
tert-Butyl-3-(hydroxymethyl)-2-methyl-1H-indole-1-carboxylate

Hazard: Irritant

Formula: C₁₅H₁₉NO₃
MW: 261.3163 g/mol
MDL Number: MFCD05864723
CAS Number: 914349-13-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR