tert-Butyl-4-hydroxy-3-(hydroxymethyl)-1H-indole-1-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: tert-Butyl 4-hydroxy-3-(hydroxymethyl)-1H-indole-1-carboxylate

071739-1G 071739-5G 071739-10G
102521-054EA 355.38 USD
102521-054 102521-056 102521-058
tert-Butyl-4-hydroxy-3-(hydroxymethyl)-1H-indole-1-carboxylate ≥95%
tert-Butyl-4-hydroxy-3-(hydroxymethyl)-1H-indole-1-carboxylate

Hazard: Irritant

Formula: C₁₄H₁₇NO₄
MW: 263.2900 g/mol
MDL Number: MFCD05864722
CAS Number: 914349-12-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR