4-Hydroxy-1H-indole-3-carbaldehyde ≥95%

Supplier: Matrix Scientific

071738-1G 071738-5G 071738-10G
102521-048EA 355.38 USD
102521-048 102521-050 102521-052
4-Hydroxy-1H-indole-3-carbaldehyde ≥95%
4-Hydroxy-1H-indole-3-carbaldehyde

Hazard: Irritant

Formula: C₉H₇NO₂
MW: 161.1574 g/mol
MDL Number: MFCD05864721
CAS Number: 81779-27-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR