tert-Butyl-5-cyano-3-(hydroxymethyl)-1H-indole-1-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: tert-Butyl 5-cyano-3-(hydroxymethyl)-1H-indole-1-carboxylate

071736-1G 071736-5G 071736-10G
102521-036EA 178.46 USD
102521-036 102521-038 102521-040
tert-Butyl-5-cyano-3-(hydroxymethyl)-1H-indole-1-carboxylate ≥95%
tert-Butyl-5-cyano-3-(hydroxymethyl)-1H-indole-1-carboxylate

Hazard: Irritant

Formula: C₁₅H₁₆N₂O₃
MW: 272.3000 g/mol
MDL Number: MFCD05864719
CAS Number: 914349-11-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR