1-Boc-4-benzyloxy-3-hydroxymethylindole ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Benzyloxy-3-hydroxymethylindole, N-BOC protected

071734-1G 071734-5G 071734-10G
102521-024EA 355.38 USD
102521-024 102521-026 102521-028
1-Boc-4-benzyloxy-3-hydroxymethylindole ≥95%
1-Boc-4-benzyloxy-3-hydroxymethylindole

Hazard: Irritant

Formula: C₂₁H₂₃NO₄
CAS Number: 914349-09-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR