tert-Butyl-3-(hydroxymethyl)-6-methoxy-1H-indole-1-carboxylate ≥95%

Supplier: Matrix Scientific

071731-1G 071731-5G 071731-10G
102521-012EA 355.38 USD
102521-012 102521-014 102521-016
tert-Butyl-3-(hydroxymethyl)-6-methoxy-1H-indole-1-carboxylate ≥95%
tert-Butyl-3-(hydroxymethyl)-6-methoxy-1H-indole-1-carboxylate

Hazard: Irritant

Formula: C₁₅H₁₉NO₄
CAS Number: 914349-08-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR