1-Boc-3-formyl-6-methoxyindole ≥95%

Supplier: Matrix Scientific

071730-1G 071730-5G 071730-10G
102521-006EA 355.38 USD
102521-006 102521-008 102521-010
1-Boc-3-formyl-6-methoxyindole ≥95%
1-Boc-3-formyl-6-methoxyindole

Hazard: Irritant

Formula: C₁₅H₁₇NO₄
CAS Number: 847448-73-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR