tert-Butyl-3-(hydroxymethyl)-5-nitro-1H-indole-1-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: tert-Butyl 3-(hydroxymethyl)-5-nitro-1H-indole-1-carboxylate, 3-(Hydroxymethyl)-5-nitro-1H-indole, N-BOC protected

071729-1G 071729-5G 071729-10G
102521-000EA 355.38 USD
102521-000 102521-002 102521-004
tert-Butyl-3-(hydroxymethyl)-5-nitro-1H-indole-1-carboxylate ≥95%
tert-Butyl-3-(hydroxymethyl)-5-nitro-1H-indole-1-carboxylate

Hazard: Irritant

Formula: C₁₄H₁₆N₂O₅
MW: 292.2872 g/mol
MDL Number: MFCD05864712
CAS Number: 914349-07-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR