tert-Butyl-3-formyl-5-nitro-1H-indole-1-carboxylate ≥95%

Supplier: Matrix Scientific

071728-1G 071728-5G 071728-10G
102520-994EA 355.38 USD
102520-994 102520-996 102520-998
tert-Butyl-3-formyl-5-nitro-1H-indole-1-carboxylate ≥95%
tert-Butyl-3-formyl-5-nitro-1H-indole-1-carboxylate

Hazard: Irritant

Formula: C₁₄H₁₄N₂O₅
CAS Number: 914349-06-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR