tert-Butyl-5-bromo-3-(hydroxymethyl)-1H-indole-1-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Bromo-3-(hydroxymethyl)-1H-indole, N-BOC protected, tert-Butyl 5-bromo-3-(hydroxymethyl)-1H-indole-1-carboxylate

071726-1G 071726-5G 071726-10G
102520-982EA 249.23 USD
102520-982 102520-984 102520-986
tert-Butyl-5-bromo-3-(hydroxymethyl)-1H-indole-1-carboxylate ≥95%
tert-Butyl-5-bromo-3-(hydroxymethyl)-1H-indole-1-carboxylate

Hazard: Irritant

Formula: C₁₄H₁₆BrNO₃
MW: 326.1857 g/mol
MDL Number: MFCD05864710
CAS Number: 905710-14-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR