tert-Butyl-3-(hydroxymethyl)-5-methyl-1H-indole-1-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: tert-Butyl 3-(hydroxymethyl)-5-methyl-1H-indole-1-carboxylate

071723-1G 071723-5G 071723-10G
102520-964EA 355.38 USD
102520-964 102520-966 102520-968
tert-Butyl-3-(hydroxymethyl)-5-methyl-1H-indole-1-carboxylate ≥95%
tert-Butyl-3-(hydroxymethyl)-5-methyl-1H-indole-1-carboxylate

Hazard: Irritant

Formula: C₁₅H₁₉NO₃
MW: 261.3163 g/mol
MDL Number: MFCD05864707
CAS Number: 914349-03-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR