tert-Butyl-4-chloro-3-(hydroxymethyl)-1H-indole-1-carboxylate ≥95%

Supplier: Matrix Scientific

071722-1G 071722-5G 071722-10G
102520-958EA 355.38 USD
102520-958 102520-960 102520-962
tert-Butyl-4-chloro-3-(hydroxymethyl)-1H-indole-1-carboxylate ≥95%
tert-Butyl-4-chloro-3-(hydroxymethyl)-1H-indole-1-carboxylate

Hazard: Irritant

Formula: C₁₄H₁₆ClNO₃
CAS Number: 914349-01-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR