tert-Butyl-4-chloro-3-formyl-1H-indole-1-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Chloro-3-formyl-1H-indole, N-BOC protected

071721-1G 071721-5G 071721-10G
102520-952EA 355.38 USD
102520-952 102520-954 102520-956
tert-Butyl-4-chloro-3-formyl-1H-indole-1-carboxylate ≥95%
tert-Butyl-4-chloro-3-formyl-1H-indole-1-carboxylate

Hazard: Irritant

Formula: C₁₄H₁₄ClNO₃
CAS Number: 914349-00-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR