4-Chloroindole-3-carbaldehyde ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Chloro-1H-indole-3-carbaldehyde

071720-1G 071720-5G 071720-10G
102520-946EA 75.38 USD
102520-946 102520-948 102520-950
4-Chloroindole-3-carbaldehyde ≥95%
4-Chloroindole-3-carbaldehyde

Hazard: Irritant

Formula: C₉H₆ClNO
MW: 179.6030 g/mol
MDL Number: MFCD05864704
CAS Number: 876-72-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR